Learn More
Agilent BioTek Filter Cubes for Imaging Multi-Mode Reader
For use with Imaging Multi-Mode Readers.
4355.00CHF
Product Code | Brand | Label or Dye | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Label or Dye | Price | Quantity & Availability | |||||
15869294
![]() |
Agilent BioTek
1225111 |
Propidium Iodide |
4355.00CHF
Each |
Estimated Shipment: 23-06-2025
Log in to see stock. |
|||||
16532082
![]() |
Agilent BioTek
1225112 |
Chlorophyll |
4355.00CHF
Each |
Estimated Shipment: 23-06-2025
Log in to see stock. |
|||||
16542082
![]() |
Agilent BioTek
1225113 |
Phycoerthyrin |
4355.00CHF
Each |
Estimated Shipment: 23-06-2025
Log in to see stock. |
|||||
16552082
![]() |
Agilent BioTek
1225114 |
Cy5.5 |
4355.00CHF
Each |
Estimated Shipment: 23-06-2025
Log in to see stock. |
|||||
16522082
![]() |
Agilent BioTek
1225110 |
CFP-YFP FRET |
4355.00CHF
Each |
Estimated Shipment: 24-07-2025
Log in to see stock. |
|||||
16512082
![]() |
Agilent BioTek
1225109 |
ACR OR |
4355.00CHF
Each |
Estimated Shipment: 23-06-2025
Log in to see stock. |
|||||
Description
For use with Imaging Multi-Mode Readers.Order Info
Requires LED.
Specifications
Filter Cube | |
Imaging Multi-Mode Reader |
Spot an opportunity for improvement?Share a Content Correction