Learn More
AIMS™ ATS-3 General Rodent Tattoo System Needles
Developed to minimize animal pain and tissue damage during tattooing
Brand:  AIMS™ 3035
Encompass
 View more versions of this product
Product Code. 13498789
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
This item is not returnable. View return policy
Description
Description
Accessory for ATS-3 General Rodent Tattoo System
  • Made of 401 stainless steel and silver solder
  • Pre-sterilized and ready for tattooing
  • Proper needles allows for humane and rapid animal tattooing
  • Tails are not damaged
  • 5 needles per pack
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.
Specifications
Specifications
Tattooing
ATS-3 general rodent tattoo system
Sterile
Tattoo Needle
401 Stainless Steel, Silver Solder
Ultra Fine Point
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.