Learn More
Ansell AlphaTec™ 37-210 Reusable Nitrile Gloves with UltraGrip™ Technology
UltraGrip™ Technology delivers an innovative raised diamond grip on the glove's surface for enhanced grip capabilities in wet and oily conditions.
Brand: Ansell 37210120
This item is not returnable.
View return policy
Description
- Reusable nitrile gloves designed specifically for food processing and handling
- Raised diamond-pattern enhances grip in wet and oily conditions
- Gloves maintain flexibility and dexterity, even at low temperatures
- Use alone or as outer glove over thermal liners or cut-resistant gloves
- Latex-free formulation presents no risk of Type 1 allergies
Specifications
ANSI 4 | |
Blue | |
Specific | |
Nitrile | |
8 mil | |
Type B | |
37-210 |
EN 420:2003 + A1:2009, REACH Compliant | |
Reusable | |
Yes | |
12 | |
3001X | |
JKT |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction