Learn More
Sigma Laborzentrifugen™ Angle Rotors for Sigma™ Series 4 and 6 Centrifuges
Use these rotors with maximum load and at maximum speed for up to 50,000 cycles with low maintenance. Sigma™ Angle Rotors for Sigma™ Series 4 and 6 Centrifuges are ideal for use in research laboratories.
Brand: Sigma Laborzentrifugen™ 12130
This item is not returnable.
View return policy
Description
- Fixed-angle rotors are used for sedimentation with high centrifugal forces of up to and more than 60,000 x g
- Allow shorter sedimentation times in comparison to swing-bucket rotors
- Higher rotational speeds and higher centrifugal forces can be achieved
- Sedimentation distance is also generally shorter than with swing-bucket rotors
- Capacity of the rotor chamber can be better utilized than with swing-bucket rotors
- Rotors can be operated continuously at maximum speed and +4°C in nearly all Sigma centrifuges
Specifications
Angle Rotor | |
13,500 rpm | |
6.7 cm | |
45° |
30 x 1.5/2.2 mL | |
10 cm | |
Sigma 4-16, 4-16K, 4-16KH, 4-16KR, 6-16, 6-16K |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction