Learn More
Anti-Human TNFRSF9/CD137 (4B4-1), Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 65159-1-IG-100UG
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The TNFRSF9/CD137 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant human cd137 ectodomain of human TNFRSF9/CD137. This antibody recognizes human, non-human primates antigen. The TNFRSF9/CD137 antibody has been validated for the following applications: FC analysis.Especificaciones
TNFRSF9/CD137 | |
Monoclonal | |
1 mg/mL | |
4-1BB; CD137; CDw137; ILA; MGC2172 | |
Recombinant human CD137 ectodomain | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Flow Antibody | |
IgG1 κ |
Flow Cytometry | |
4B4-1 | |
Unconjugated | |
Mouse | |
Purified by protein-A affinity chromatography | |
RUO | |
3604 | |
Store at 2-8°C. Stable for one year after shipment. | |