Learn More
BD Microlance 3 Special Cannulas
Click to view available options
Needle Gauge:
21 G
Unit Size:
Pack of 100
Description
- Extremely thin walls enable high flow rates
- Combined Luer-/BD-Luer-Lock adapter
- Transparent, colour coded (green) plastic adapter
- Packaged in 100 single use packs to maintain sterility
Specifications
Specifications
Material | Silicon-coated |
Color | Green Adapter |
Description | Microlance |
Needle Gauge | 21 G |
Diameter (Metric) Outer | 0.81 mm |
Includes | Luer/Luer-Lock adapter |
Latex Free | Yes |
Length (English) Needle | 1 in. |
Product Type | Thin wall Hypodermic Needle |
Needle Point Style | Triple Facet, Short Bevel |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction