Learn More
Proteintech Broad Range Prestained Protein Marker
The PL00002 Broad Range Protein Marker is a ready-to-use three-color protein standard with 13 prestained proteins covering a wide range of molecular weights from 3 to 245 kDa in Tris-Glycine buffer.
Marca: Proteintech PL00002-100UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- The PL00002 Broad Range Prestained Protein Marker is designed for monitoring protein separation during SDS-polyacrylamide gel electrophoresis, verification of Western transfer efficiency on membranes (nitrocellulose, PVDF, or nylon) and for estimating the size of proteins.
- Contents: Approximately 0.1~0.4 mg/mL of each protein in the buffer (20 mM Tris-phosphate (pH 7.5 at 25°C), 2 % SDS, 0.2 mM Dithiothreitol, 3.6 M Urea, and 15 % (v/v) Glycerol).
Especificaciones
Colorimetric | |
3 to 245 | |
100 Tests |
13 | |
Blue, Red, Green | |
Aliquot and store at -20™C for up to 1 year. |