Learn More
Unity Lab Services™ Calibration Certificate Cofrac Calipers
ULS offers a full range of support services and programs for all instruments and supplies, regardless of manufacturer. Maximize your technology investments by anticipating and avoiding problems before they occur.
Brand: Unity Lab Services™ UMV-P9361
This item is not returnable.
View return policy
Description
- Visual and manual examination of zero control, adjustment of slide clearance, estimation of the flatness of the measuring faces, measurement of the indication error (2 measurements in 6 positions distributed over the range), analysis of the results to standard XP E11-091, provision of a COFRAC calibration certificate and a verification report
- Valid for calipers with a length less than or equal to 300mm
- Other caliper lengths and timelines: contact us
Specifications
Calipers | |
Calibration Certificate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction