Learn More
Unity Lab Services™ Calibration Certificate Cofrac Temperature Two Fixed Points, 0°C, 4°C
ULS offers a full range of support services and programs for all instruments and supplies, regardless of manufacturer. Maximize your technology investments by anticipating and avoiding problems before they occur.
Brand:  Unity Lab Services™ CAL935E
Product Code. 13475829
  • POA / Each
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Cet article nécessite une mise à jour du prix avant d'être commandé.
Demandez un prix pour cet article ici.
View alternative products »
This item is not returnable. View return policy
Description
Description
  • Calibration in comparison with a reference thermometer in baths
  • Type of device: temperature measuring thermometric chain or expansion thermometer at fixed values ​​of: 0°C and 4°C
  • Our best uncertainties for a thermometer with a resolution of 0.01°C (k = 2): +/- 0.02°C to 0°C; +/- 0.05°C to 4°C
  • Our best uncertainties for a thermometer with a resolution of 0.1 ° C (k = 2): +/- 0.1°C to 0°C; +/- 0.2°C to 4°C
  • COFRAC Accreditation Number 2-5984
  • Calibration, scope available on www.cofrac.fr
  • Average time: 1 week
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.