missing translation for 'onlineSavingsMsg'
Learn More

CALR3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Especificaciones

Antígeno CALR3
Concentración 0.13 mg/mL
Aplicaciones Western Blot, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
View More Specs

Products 2
Product Code Brand Cantidad Price Quantity & Availability  
Product Code Brand Cantidad Price Quantity & Availability  
16816894
View Documents
Proteintech
17064-1-AP-20UL
20 μL
145.00€
20 microlitros
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16806894
View Documents
Proteintech
17064-1-AP-150UL
150 μL
410.00€
150 microlitros
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description

Description

Calreticulins, such as Calreticulin 3, are Ca(2+)-binding chaperones localized mainly in the endoplasmic/sarcoplasmic reticulum.
Specifications

Specifications

CALR3
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Mouse, Human, Rat
Q96L12, Q9D9Q6
125972, 364529, 73316
CALR3 Fusion Protein Ag10847
Primary
-20°C
CALR3
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CALR3, Calreticulin 2, calreticulin 3, CRT2
Calr3
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.