Learn More
behr Labor-Technik™ HDPE, Canister with Screw Cap and UN Approval
Click to view available options
Capacity (Metric):
5 L
10 L
20 L
25 L
30 L
60 L
Description
- White High-Density Polyethylene canisters
- UN-certified
- Screw-on cap
- Suitable for hazardous goods
Specifications
Specifications
Capacity (English) | 7.9 gal. |
Capacity (Metric) | 30 L |
Certifications/Compliance | UN-certified |
Closure Material | Polyethylene |
Closure Type | Screw Cap |
Color | White |
Description | PE canister 30 L, white |
For Use With (Application) | For Transport and Storage of Hazardous Goods |
Includes | Screw cap |
Material | HDPE |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction