Learn More
CD127 (IL-7Ra) Rat anti-Mouse, PE, Clone: A7R34, Proteintech
Rat Monoclonal Antibody
Marca: Proteintech PE-65093-100UG
Descripción
The CD127 (IL-7Ra) antibody from Proteintech is a rat monoclonal antibody generated with il-7ra-igg1 fusion protein of mouse CD127 (IL-7Ra). This antibody recognizes mouse antigen. The CD127 (IL-7Ra) antibody has been validated for the following applications: FC analysis. This antibody is pe.
Especificaciones
CD127 (IL-7Ra) | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
Primary | |
Mouse |
Flow Cytometry | |
A7R34 | |
PE | |
100 μg | |
16197 | |
IgG2a |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.