missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD336 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 55118-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The antibody is specific to CD336.
CD336 is also known as NKp44 or NCR2. CD336 is a 44-kDa transmembrane glycoprotein belonging to the natural cytotoxicity receptor (NCR) family and contains an ITIM domain located in the cytoplasmic tail. Expression of CD336 and NK cell activation by the receptor requires association with DAP12, an ITAM-containing transmembrane accessory protein. Once thought to be expressed exclusively on IL-2-activated NK cells and a subset of gamma delta T lymphocytes, studies have shown expression of CD336 on IL-3-stimulated plasmacytoid dendritic cells, alpha beta T cells in the small intestinal epithelium, and T cells in cord blood. CD336 has been shown to be involved in the innate response of activated NK cells against viral infection and tumor cell proliferation.Especificaciones
CD336 | |
Polyclonal | |
Unconjugated | |
NCR2 | |
CD336, LY95, Lymphocyte antigen 95 homolog, NCR2, NK cell activating receptor, NK p44, NKP44 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9436 | |
-20°C | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95944 | |
NCR2 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido