Learn More
CD4 Rat anti-Mouse, APC, Clone: RM4-5, Proteintech
Rat Monoclonal Antibody
Marca: Proteintech APC-65141-100UG
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The CD4 antibody from Proteintech is a rat monoclonal antibody generated with balb/c mouse thymocytes of mouse CD4. This antibody recognizes mouse antigen. The CD4 antibody has been validated for the following applications: FC analysis. This antibody is apc.
Especificaciones
CD4 | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
Primary | |
Mouse |
Flow Cytometry | |
RM4-5 | |
APC | |
100 μg | |
12504 | |
IgG2a |