Learn More
CD45.2 Mouse anti-Mouse, CoraLite 647, Clone: 104, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL647-65072-100TESTS
Descripción
The CD45.2 antibody from Proteintech is a mouse monoclonal antibody generated with b10.s mouse thymocytes and splenocytes of mouse CD45.2. This antibody recognizes mouse antigen. The CD45.2 antibody has been validated for the following applications: FC analysis. This antibody is coralite™647.
Especificaciones
CD45.2 | |
Monoclonal | |
5 μL/Test | |
Mouse | |
Primary | |
Mouse |
Flow Cytometry | |
104 | |
CoraLite 647 | |
100 Tests | |
19264 | |
IgG2a |