Learn More
Conductivity Calibration Solution, 445μS, Cole-Parmer™
Brand:  Cole Parmer 01489-93
Quantity 946 mL
Product Code. 15262046
  • 54.60€ / Each
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
946mL
214.3ppm Nacl
946 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.