Learn More
CSTF1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15537-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes one of three subunits which combine to form cleavage stimulation factor (CSTF). CSTF is involved in the polyadenylation and 3′end cleavage of pre-mRNAs. Similar to mammalian G protein beta subunits, this protein contains transducin-like repeats. Several transcript variants with different 5′ UTR, but encoding the same protein, have been found for this gene.Especificaciones
CSTF1 | |
Polyclonal | |
Unconjugated | |
CSTF1 | |
CSTF1 | |
CSTF1 Fusion Protein Ag7874 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q05048 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1477 | |
-20°C | |