Learn More
Amarell™ Fine-Graduation Hydrometer without Thermometer, series M100
Calibrated for reference temperature 20°C. The high-graduation density hydrometer without thermometer is DIN rated for many specific gravity measurements.
Brand:  Amarell™ H809606
 View more versions of this product
Product Code. 10503182
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Reference temperature 20°C
  • Each hydrometer length is 250mm
Specifications
Specifications
1Each
±1g/cm3
250 mm
20°C
Density Hydrometer
0.0020 g/cm3
0.900 to1.000 g/cm3
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.