Learn More
BUCHI Drying Rack Manifold 24
Connect up to 24 samples to a BUCHI Lyovapor™. Has 24 manifold valves and a base plate.
Marca:  BUCHI 11DCIN02000
4860.95 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.
Código de producto. 15962701
  • 4990.00€ / cada uno
Solo quedan null
Fecha estimada de envoi: 04-11-2025
Descripción
  • Manifolds: adapters for traditional freeze drying beakers or round bottom flasks
  • Shelves: for vials, plates, and irregular shaped items
  • Heated Shelves: advanced control of drying process and applications
  • Stoppering Chambers: seal vials without exposure to ambient moisture
Especificaciones
Especificaciones
Benchtop Freeze Dryer
Ambient to -105°C
North America
0
Acero inoxidable
220/240 V