Learn More
MilliporeSigma™ Silica Gel 60G TLC Plates, Glass Backed
These new TLC silica gel 60G plates are recommended for customers in QA/QC labs using older Ph Eur monograph methods, which require TLC plates with gypsum binder.
Brand: MilliporeSigma™ 1.00390.0001
Description
- Silica gel coated with flourescent indicator F254
- 20 x 20cm
- Provided in packs of 25
- For thin layer chromatography applications
Specifications
25/Pk. | |
F254 | |
20 x 20 cm |
TLC Silica Gel 60 G F254 | |
250 μm |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.