Learn More
EXO1 Mouse, anti-Human, Clone: 2D11D1, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 68536-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The EXO1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EXO1. This antibody recognizes human antigen. The EXO1 antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
EXO1 | |
2D11D1 | |
EXO1, EXOI, exonuclease 1, Exonuclease I, HEX1, hExo1, hExoI | |
150 μL | |
9156 | |
Store at -20°C. Stable for one year after shipment. | |
Liquid |
Western Blot, ELISA | |
1000 μg/mL | |
Mouse | |
Primary | |
Human | |
150 kDa | |
IgG1 |
Título del producto
EXO1 Mouse, anti-Human, Clone: 2D11D1, Proteintech > Quantity: 150 μL
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido