Learn More
Fascin Mouse anti-Human, CoraLite 488, Clone: 1B5G3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-66321-100UL
Descripción
The Fascin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Fascin. This antibody recognizes human antigen. The Fascin antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Especificaciones
Fascin | |
Monoclonal | |
CoraLite 488 | |
100 μL | |
6624 | |
IgG2a |
Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
1B5G3 | |
Mouse | |
Primary | |
Human |