Learn More
BUCHI FlashPure Empty Solid Loader: Parts
Parts for FlashPure Empty Solid Loaders. FlashPure Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatography. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
98.38€ - 155.00€
Product Code | Brand | Includes | Type | Size (Metric) Tube Rack | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Type | Size (Metric) Tube Rack | Price | Quantity & Availability | |||
16360316
|
BUCHI
148634551 |
20 Pc. | Frit | 12 g |
105.00€
Pack of 20 |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||
16380316
|
BUCHI
148634560 |
10 Pc. | Frit | 40 g |
98.38€
Pack of 10 |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||
16340276
|
BUCHI
142129062 |
1 Pc. | Insertion Rod | 12 to 20 g |
126.00€
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||
16330276
|
BUCHI
142129061 |
1 Pc. | Insertion Rod | 20 to 40 g |
144.56€
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||
16300276
|
BUCHI
140000178 |
1 pc. | Insertion Rod | 80 to120 g |
155.00€
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||
Description
- Screw cap makes sealing easy
- Various sizes for differing sample amounts
- Choose a size large enough for the sample
- Insertion Rod allows frit placement and loading of samples smaller than the entire volume
Spot an opportunity for improvement?Share a Content Correction