Learn More
3M™ Cartucho de gas y vapor, Serie 6000
Combina una excelente protección con una extraordinaria comodidad y equilibrio. El cartucho de gas y vapor 3M tiene una forma trapezoidal y funciona con máscaras faciales completas y medias que tienen sistemas de conexión en bayoneta.
Marca: 3M™ 7000034747
Descripción
Each filter has a colored band using the EN14387 classification system, to denote the nature of the protection it provides.
- Trapezoidal-shaped gas and vapor filters combine excellent protection with superb comfort and balance
- Lightweight, low breathing resistance, and well balanced when fitted to appropriate mask
- Provides excellent field of vision as a result of the unique trapezoidal shape.
- Filters can be used on 3M™ Facemasks fitted with a bayonet connections system