Learn More
GDI2 Mouse anti-Human, CoraLite 488, Clone: 4B8F3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-60078-100UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The GDI2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GDI2. This antibody recognizes human, mouse antigen. The GDI2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Especificaciones
GDI2 | |
Monoclonal | |
2000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse |
Immunofluorescence | |
4B8F3 | |
CoraLite 488 | |
100 μL | |
2665 | |
IgG1 |