Learn More
Amarell™ Standard Density Hydrometer without Thermometer, measuring span 0.200g/cm3
Calibrated for reference temperature 20°C. The standard density hydrometer without thermometer can be used for many general specific gravity measurements.
Brand:  Amarell™ H801364
 View more versions of this product
Product Code. 10130542
  • 31.20 € / Each
Estimated Shipment: 26-05-2025
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Reference temperature 20°C
  • Each hydrometer length is 280mm
Specifications
Specifications
Density Hydrometer
0.002 g/cm3
1000 to 1.200 g/cm3
±1g/cm3
280 mm
20°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.