Learn More
Vilber Lourmat™ Germicidal UV Lamp
Brand: Vilber Lourmat™ 3121-1151-1
This item is not returnable.
View return policy
Specifications
230 V | |
254 nm | |
15 W, 230 V, 50 Hz | |
50 Hz | |
31 mW/cm2 | |
Germicidal UV Lamp |
15 W | |
140 mm | |
117 mm | |
505 mm | |
1 Ea. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction