Learn More
Greiner Bio-One Serum Tubes: Clot Activator with Gel Separator
For routine clinical chemistry, immunohematology, serology or immunology testing
Merk:  Greiner Bio-One 455071
 Bekijk meerdere types van dit product via de
Productcode. 10144534
  • 243.96 /
This item is not returnable. View return policy
Beschrijving
Beschrijving
  • Coated with micronized silica particles which activate clotting when tubes are gently inverted
  • Tubes with gel contain a barrier gel on the tube bottom that during centrifugation moves upward to the serum-clot interface to form a stable barrier, separating serum from cells
  • Clear as glass with equal or better performance
  • Molded to be virtually unbreakable, providing a better level of safety than glass
  • Tubes have an 18-month shelf life from the date of manufacture
  • Pediatric tubes come with white rings for quick identification
  • Safety Twist Caps can be removed and replaced several times
  • Nonscrew caps can be removed and replaced as often as required
Compliance
Tubes with gel separator are FDA 510(k) cleared for therapeutic drug monitoring (TDM)
TRUSTED_SUSTAINABILITY
Specificatie
Specificatie
Microscopic Silica Particles with Separation Gel
Non-ridged Pull Cap
Red with Yellow Ring
Plastic
8 mL
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.