Learn More
Schotte Kunststoffen™ Natural HDPE Bottles with Wide Mouth
Use Schotte Kunststoffen™ Natural HDPE Bottles for storage and transfer of materials in the laboratory. They have a wide neck and are packaged in various quantities.
€ 202.00 - € 1099.81
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10207544
|
Schotte Kunststoffen™
10197 |
250 mL | N/A | ||||||
10358754
|
Schotte Kunststoffen™
10309 |
500 mL | N/A | ||||||
10607975
|
Schotte Kunststoffen™
208CR-55-AK1 |
1000 mL | N/A | ||||||
Description
- Recyclable
- Natural color
- Wide mouth
Specifications
White | |
HDPE | |
Wide | |
Wide Mouth Bottle |
Spot an opportunity for improvement?Share a Content Correction