Learn More
Brannan™ Hydrometer specific gravity shot loaded medium adjustment for surface tension 1.400 to 1.450 x 0.001 S.G. 270 mm Brannan
Slot loaded density hydrometer for a variety of surface tension ranges.
Brand:  Brannan™ 10/932/0
Product Code. 12891468
  • € 22.25 / Each
Only null left
Estimated Shipment: 08-08-2025
Add to basket
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
Specific Gravity Hydrometer
270 mm
Soda Glass
0.001 g/mL
1.40 to 1.45 g/mL
Steel Shot
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.