Learn More
Honeywell Safety Products™ Ignite Glasses
Click to view available options
Color de marco:
Black, Silver
Red, Silver
Tamaño de la unidad:
cada uno
paquete de 10
Tinción de lente:
Gray
Description
- Nine-base, wraparound lens design.
- Close-to-the-face protection and superior coverage.
- Lightweight frameless design.
- All-day comfort and180° of distortion free, unobstructed vision.
- Adjustable nose-piece.
- Reduced slippage and personalized, comfortable, secure fit.
Specifications
Specifications
Certificaciones/conformidad | EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1775, EC Attestation Number EC1775 |
Revestimiento de lente | Impact-resistant |
Tipo de producto | Gafas de seguridad |
Para utilizar con (aplicación) | Indoor use. Automotive, Building, Construction, Catering, Fishing, Food, Maintenance, Service and Wood industries. |
Color de marco | Red, Silver |
N.° por paquete | 1 Ea. |
Lens Material | Policarbonato |
Material del bastidor | Polyamide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction