Learn More
Honeywell Safety Products™ Ignite Glasses
Código de producto. 12970345
Click to view available options
Color de marco:
Black, Silver
Red, Silver
Tamaño de la unidad:
cada uno
paquete de 10
Tinción de lente:
Gray
Lightweight, frameless, distortion free, unobstructed vision safety glasses. Honeywell Safety Products™ Ignite Safety Glasses offer a comfortable and secure fit.
  • Nine-base, wraparound lens design.
  • Close-to-the-face protection and superior coverage.
  • Lightweight frameless design.
  • All-day comfort and180° of distortion free, unobstructed vision.
  • Adjustable nose-piece.
  • Reduced slippage and personalized, comfortable, secure fit.
TRUSTED_SUSTAINABILITY
Especificaciones
Certificaciones/conformidad EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1775, EC Attestation Number EC1775
Revestimiento de lente Impact-resistant, Hardcoat
Tipo de producto Gafas de seguridad
Para utilizar con (aplicación) Indoor use. Automotive, Building, Construction, Maintenance, Paper, Ship Building, Transportation and Utility industries.
Color de marco Black, Silver
N.° por paquete 1 Ea.
Lens Material Policarbonato
Material del bastidor Polyamide