Learn More
Cole-Parmer™ Integrity 10, RS-200, and RS-250 Reaction Station Accessories
Click to view available options
Descripción:
Multi IR box
Módulo de temperatura múltiple
PID-NIR5-BNSD
Sensor IR no intrusivo
Software de integridad
Vidrio de 24 mm × 150 mm
Descripción
- Suitable for use with Cole-Parmer™ Integrity 10, RS-200, or RS-250 series reaction stations.
- Choose from a wide range of accessories suitable for your reaction station including reaction vessels, probes, and software.
Especificaciones
Especificaciones
Descripción | PID-NIR5-BNSD |
Para utilizar con (equipo) | Integrity 10 |
Material | Acero inoxidable |
Tipo | IR Probe |