Learn More
DWK Life Sciences Kimble™ Kontes™ Adsorption/Ion-Exchange Columns
With or without fritted disc
1780.00 SEK - 2815.00 SEK
Specifications
Type | Adsorption/Ion Exchange Chromatography Column |
---|---|
For Use With (Application) | Many separations in adsorption and ion exchange chromatography |
Material | Borosilicate Glass |
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Column Capacity | Includes | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Column Capacity | Includes | Price | Quantity & Availability | ||
11887413
|
DWK Life Sciences
420320-0213 |
300 mm | 11 mm | 20 mL | Without fritted disc |
1780.00 SEK
Each |
In Stock
Log in to see stock. |
||
11807423
|
DWK Life Sciences
420320-0233 |
300 mm | 22 mm | 110 mL | Fritted disc |
2110.00 SEK
Each |
Estimated Shipment: 12-12-2025
Log in to see stock. |
||
11837423
|
DWK Life Sciences
420320-0256 |
600 mm | 41 mm | 790 mL | Fritted disc |
2815.00 SEK
Each |
Estimated Shipment: 12-12-2025
Log in to see stock. |
||
11827423
|
DWK Life Sciences
420320-0245 |
500 mm | 25 mm | 250 mL | Fritted disc |
2335.00 SEK
Each |
Estimated Shipment: 12-12-2025
Log in to see stock. |
||
11897413
|
DWK Life Sciences
420320-0224 |
400 mm | 19 mm | 110 mL | Fritted disc |
1970.00 SEK
Each |
Estimated Shipment: 12-12-2025
Log in to see stock. |
||
Description
- Borosilicate glass
- Sealed-in coarse porosity (40 to 60μm) fritted disc supports packing in K420320 Series columns
- Use glass wool to support packing in K420300 Series columns (packing not included)
Specifications
Adsorption/Ion Exchange Chromatography Column | |
Many separations in adsorption and ion exchange chromatography | |
Borosilicate Glass |
Spot an opportunity for improvement?Share a Content Correction