Learn More
Kimtech™ Pure™ A6 Clean Overalls
Maintain ISO 7 Class C or higher cleanroom environments with breathable, ultra light overalls. The Kimtech Pure™ A6 Clean Overalls are made of a high performance, chemical resistant fabric that are origami folded to reduce gowning time.
Especificaciones
Tipo de producto | Cleanroom Coveralls |
---|---|
Material | SMS |
Color | Blanco |
Desechables | Single-use |
Estilo de manguito | Thumb Loop |
Descripción
- High performance chemical resistant fabric
- Origami folded to reduce gowning time
- Presented unzipped and inside-out reducing the risk of contaminating the outside of the garment
Especificaciones
Cleanroom Coveralls | |
Blanco | |
Thumb Loop | |
No estéril | |
Breathable | |
Controlled Environments |
SMS | |
Single-use | |
Con anillo | |
Soporte | |
PPE Cat III Type 5/6 (Protection against particles/limited splash protection). Add Infectious agent protection. EN1149-1:1995 Antistatic garment |