Learn More
UVP Lamp 3Uv-34 3Uv Multi-Wavelength Supplied with 1 X 254Nm
Brand:  UVP 95-0341-02
Product Code. 12873898
  • € 1745.00 / Each
Only null left
Estimated Shipment: 01-10-2025
Add to basket
This item is not returnable. View return policy
Description
Description
1 x 302nm and 1 x 365nm tube 230V 50Hz 4W UVP
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.