Learn More
LARS2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 17097-1-AP-150UL
 Ver más versiones de este producto
Código de producto. 16806944
  • 410.00€ / 150 microlitros
Descripción
This gene encodes a class 1 aminoacyl-tRNA synthetase, mitochondrial leucyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid.
Especificaciones
Especificaciones
LARS2
Polyclonal
Unconjugated
LARS2
KIAA0028, LARS2, Leucine tRNA ligase, LEURS
Rabbit
Antigen Affinity Chromatography
RUO
23395
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q15031
LARS2
LARS2 Fusion Protein Ag10576
150 μL