Learn More
BUCHI Manifold Pressure Difference Test Set for Lyovapor™
The Lyovapor™ sensors accurately track temperature and/or pressure throughout the freeze-drying process, ensuring optimized operations and facilitating accurate endpoint determination using capacitive, Pirani pressure gauges, and sample temperature probes.
Marca:  BUCHI 11080770
3335.03 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.
Código de producto. 30130647
  • 3425.00€ / cada uno
Solo quedan null
Fecha estimada de envoi: 31-07-2025
Descripción
  • Determine endpoint completion in a manifold configuration
Especificaciones
Especificaciones
Benchtop Freeze Dryer
0
Acero inoxidable
220/240 V
4.5 L
Ambient to -105°C
North America