missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRE11A Mouse anti-Human, Unconjugated, Clone: 1B12B2, Proteintech
Mouse Monoclonal Antibody
1081.84 DKK - 3058.99 DKK
Specifications
Antigen | MRE11A |
---|---|
Clone | 1B12B2 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16860276
|
Proteintech
67570-1-IG-20UL |
20 μL |
1081.84 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16850276
|
Proteintech
67570-1-IG-150UL |
150 μL |
3058.99 DKK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MRE11A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MRE11A. This antibody recognizes human, mouse, rat antigen. The MRE11A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
MRE11A | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
4361 | |
Primary |
1B12B2 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title