missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYOZ1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13160-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The MW of this protein is 32 kDa, and 13160-1-AP specifically recognizes the 32 kDa protein.
Sarcomere assembly is regulated by the muscle protein titin. Titin is a giant elastic protein with kinase activity that extends half the length of a sarcomere. It serves as a scaffold to which myofibrils and other muscle related proteins are attached. This gene encodes a protein found in striated and cardiac muscle that binds to the titin Z1-Z2 domains and is a substrate of titin kinase, interactions thought to be critical to sarcomere assembly. Mutations in this gene are associated with limb-girdle muscular dystrophy type 2G.Especificaciones
MYOZ1 | |
Polyclonal | |
Unconjugated | |
MYOZ1 | |
Calsarcin 2, CS 2, FATZ, MYOZ, MYOZ1, myozenin 1, Protein FATZ | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
498440, 58529, 59011 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9JK37, Q9NP98 | |
Myoz1 | |
MYOZ1 Fusion Protein Ag3806 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido