Learn More
Kinesis™ Omnifit™ Gripper Fittings
Flangeless systems with 1/4 in.-28 United National Fine (UNF) thread provides connection for tubing with an outside diameter of 1/16 in. or 1/8 in.
Specifications
Type Gripper Fitting
Max. Pressure 1000 psi
View More Specs
Description
Description
  • Incorporate a PTFE-seal press-fitted into 316 stainless-steel housing for leak-resistant connection with minimal disturbance to the fluid path
  • Allow repeated connect/disconnect without sealing-face deterioration
  • Gripper fittings do not compress the tubing if over-tightened, and do not twist the tubing during connection
  • Pressure rated to 1000 psi (68 bar)
Use with Omnifit Tube End Fittings
To Use
  • Trim tubing to a point and slide on Tube End fitting
  • Add the Gripper fitting to tubing, ensuring the PTFE seal faces the pointed tubing end
  • Using pliers, pull tubing through Gripper fitting until PTFE seal has reached the uncut portion
  • Rotate Gripper fitting around tubing to seat; trim excess tubing off
Specifications
Specifications
Gripper Fitting
1000 psi
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.