Learn More
Pepsin/Hydrochloric Acid Electrode Cleaning Solution, SI Analytics™
SI Analytics™ Pepsin/Hydrochloric Acid Electrode Cleaning Solution is a corrosive solution that is suitable for cleaning electrodes.
404.00 NOK - 872.00 NOK
Specifications
Boiling Point | 100°C |
---|---|
Color | Colorless |
Filling Solution Type | Cleaning Solution |
Odor | Odorless |
pH Range | 1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11904759
|
SI Analytics
285138295 |
250 mL |
404.00 NOK
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10213511
|
SI Analytics
285138538 |
1000 mL |
872.00 NOK
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Water, pepsin A, hydrochloric acid
- Classification according to Regulation (EC) No 1272/2008: HS05 corrosion
- The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
- Avoid contact with the eyes and skin
- Do not inhale gases / fumes / aerosols
- Take off contaminated clothing and wash it before reuse
- Formation of hydrogen possible with metals and alloys (risk of explosion)
- Generally not hazardous for water
Specifications
100°C | |
Cleaning Solution | |
1 | |
23hPa |
Colorless | |
Odorless | |
Fully miscible in water |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Pepsin/hydrochloric acid solution
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P234-Keep only in original container.
Supplemental information
- MIXTURE LIST-Contains : Pepsin A
Spot an opportunity for improvement?Share a Content Correction