Learn More
Knick Elektronische™ Portamess™ 913 pH Advanced pH Meter
Includes datalogger and print functionalities. Smooth membrane keypad is durable and easy to clean. No sticking or crusty deposits.
Brand:  Knick Elektronische™ 913PH
Product Code. 15790528
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
This item is not returnable. View return policy
Description
Description
  • Portamess™ 913 pH Advanced pH Meter with datalogger and print functionalities includes remote-control capability.
  • Smooth membrane keypad is durable and easy to clean. No sticking or crusty deposits.
  • 15mm character height LC display with antireflection natural glass is scratchproof and resistant to chemicals.
  • Clear icons for operator guidance.
  • Integrated, replaceable quiver protects the sensor. Transparent. Easy to remove.
  • Blue, ergonomic enclosure is resistant to impact and chemicals. Water-tight, diecast chassis. (IP 66 protection).
  • Cover protects against dirt and damage.
  • Highly flexible lamellar joint can be folded back completely.
  • Fold-out for hanging or standing up keeps both hands free for handling the sensor.
  • Robust, gold-plated sockets are water-tight.
  • Adjustable carrying strap is fixed directly to the meter.
  • 1-, 2-, and 3-point calibration.
  • Calimatic™ automatically recognizes the right buffer fro 8 buffer sets.
  • Manual calibration possible with any two buffers.
  • Automatic recording of measured values over longer periods. Manual (with STO key), interval (e. g. every 30 min.) or eventcontrolled (based on a measured value difference – this function prevents unnecessary memory consumption).
  • Combined with a PC, the meter can continuously record any amount of data. Either for printer or PC simply by turning plug around. Saves paperwork, prevents manual transcription errors. Supplies records for QM documentation (ISO 9000 and GLP).
  • Paraly™ SW 109 transfer software transfers measured values to the PC.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.