Learn More
PIP5K1A Mouse anti-Human, Unconjugated, Clone: 3E5G12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67483-1-IG-150UL
This item is not returnable.
View return policy
Description
The PIP5K1A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PIP5K1A. This antibody recognizes human antigen. The PIP5K1A antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
PIP5K1A | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
3E5G12 | |
Unconjugated | |
150 μL | |
8394 | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction