missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ KleenGuard™ A80 Chemical Permeation and Liquid Protection Coveralls
Specially designed for increased protection against chemical splashes. Kimberly-Clark™ KLEENGUARD™ A80 Chemical Permeation and Jet Liquid Protection Coveralls have welded seams that improve safety in high-risk areas creating a jet-tight barrier against harmful substances.
Especificaciones
Tipo de producto | General Purpose Coveralls |
---|---|
Material | Spunbond Polypropylene |
Color | Orange |
Desechables | Limited-use |
Esterilidad | No estéril |
Descripción
- Approved as Complex Design (Category 3) equipment - offering protection to the levels specified for Type 3 and 5 by CEN
- Highly visible, single-use protective clothing to improve safety and comfort in high-risk areas - creates a high pressure jet tight barrier against harmful chemical splashes, liquids or dust particles
- Suitable for use as protection against Infective Agents and Biohazards - EN 14126
- Suitable for use as protection against solid particles and particulate radioactive contamination - EN1073-2
- Treated to be antistatic to EN 1149-5
- Ideal for working with hazardous chemicals - protecting against liquid, aerosol and dry chemical splashes, sprays, high pressure jets, fibres and dust when used for heavy industrial clean-up, maintenance, tank cleaning, petrochemical and chemical handling
- Perfect for environments where high levels of body protection are vital, such as chemical and manufacturing industries
- Durable SMS fabric with additional treatment for increased protection against chemical splashes
- Offers complete freedom of movement with comfortable body design
Especificaciones
General Purpose Coveralls | |
Orange | |
No estéril | |
Regular |
Spunbond Polypropylene | |
Limited-use | |
Sí |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto