Learn More
Thermo Scientific™ Precision™ Shaking Water Baths, Precision SWB 27 - PROMO
Shaking water baths support a range of sensitive life science and QA/QC applications, from warming fragile reagents to tissue culturing and genetics sequencing.
Marca: Thermo Scientific™ TSSWB27PMO3
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Easily clean and maintain bath with coil-free design
- Optimize scheduling with auto-on and auto-off timers
- Help prevent bath overheating and damage with low-fluid protection
- Conveniently save commonly used settings with four temperature and shaking speed presets
- Help protect your work with audible alarms
- Easily operate and monitor with icon-based graphical display
- Accommodate taller labware with new hinged lid and extended height
- Features next-generation shaker motor