Learn More
Systec™ Radial Ventilator DV Series and H-210 TO H-1580 Autoclaves
Systec™ Radial Ventilator DV Series and H-210 TO H-1580 Autoclaves facilitates air circulation and accelerated heat removal from the chamber.
Brand: Systec™ 9511
This item is not returnable.
View return policy
Description
- In conjunction with optional mantle cooling with cooling water and support pressure, the radial ventilator ensures accelerated removal of heat from the sterilization items to the cooled chamber mantle.
- The radial ventilator is located in the lid of the chamber and is driven by a magnetic motor fitted outside under the cover.
- The radial ventilator is placed in the door of the sterilization chamber so that the usable space in the autoclave is not reduced.
- Ventilation performance 250 m3/h
- Reduction of cooling time by up to 70%
Specifications
Reduces cooling time by up to 70% | |
Accelerated Autoclave Cooling | |
DV Series, H-210 to H-1580 Autoclaves | |
Radial Ventilator |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction