Learn More
SCAT Replacement Lid for Funnel MARCO V2.0 - Conductive
Replacement lid for funnel MARCO, V2.0, black, electrostatic conductive
Brand: SCAT 318988
358.01 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Replacement lid
- For funnel MARCO, V2.0
- Black
- Electrostatic conductive
Specifications
Replacement Lid | |
HDPE | |
Blue/White | |
140 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction