Learn More
Epredia™ Decalcifying Solution
Remove calcium from histological specimens with Epredia™ Decalcifying Solution.
79.00€
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12697766
|
Epredia™
8340 |
32 oz. | N/A | ||||||
Description
- Offers rapid decalcification without destroying nuclear properties of the tissue
- The EDTA and dilute hydrochloric acid in this solution require no heat, electricity, or other aids to decalcify bone tissue
Specifications
Decalcifying Solution |
Spot an opportunity for improvement?Share a Content Correction