Learn More
RREB1-Specific Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20280-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The RREB1-Specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human RREB1-Specific. This antibody recognizes human antigen. The RREB1-Specific antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Especificaciones
RREB1-Specific | |
Polyclonal | |
Unconjugated | |
20 μL | |
6239 | |
IgG |
Western Blot, Immunohistochemistry | |
187 μg/mL | |
Rabbit | |
Primary | |
Human |